Learn More
Invitrogen™ EYA1 Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Marke:  Invitrogen™ PA5117158
Artikelnummer. 17289653
  • 538.00 CHF / 100 Mikroliter
om dit product te kopen Registreer vandaag om een webaccount aan te maken
Beschreibung
Beschreibung
Positive Samples: SH-SY5Y, 293T, Mouse testis, Mouse brain, Rat brain Immunogen sequence: NSLTNSSGFN SSQQDYPSYP SFGQGQYAQY YNSSPYPAHY MTSSNTSPTT PSTNATYQLQ EPPSGITSQA VTDPTAEYST IHSPSTPIKD SDSDRLRRGS D.
This gene encodes a member of the eyes absent family of proteins. The encoded protein may play a role in the developing kidney, branchial arches, eye, and ear. Mutations of this gene have been associated with branchiootorenal dysplasia syndrome, branchiootic syndrome, and sporadic cases of congenital cataracts and ocular anterior segment anomalies. A similar protein in mice can act as a transcriptional activator. Four transcript variants encoding three distinct isoforms have been identified for this gene.
TRUSTED_SUSTAINABILITY
Spezifikation
Spezifikation
EYA1
Polyclonal
Unconjugated
EYA1
BOP; bor; BOS1; dog eared; EYA transcriptional coactivator and phosphatase 1; eya1; eya1 protein; Eyes absent; eyes absent 1; eyes absent 1 homolog; eyes absent 1 homolog (Drosophila); eyes absent homolog 1; MGC141875; OFC1; OTTHUMP00000195053; wu:fc13c10; zgc:100770
Rabbit
Affinity chromatography
RUO
14048, 2138, 502935
-20°C, Avoid Freeze/Thaw Cycles
Liquid
ELISA, Immunohistochemistry (Paraffin), Western Blot
1.85 mg/mL
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
P97767, Q99502
EYA1
A synthetic peptide corresponding to a sequence within amino acids 200-300 of human EYA1 (NP_0004942).
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Produktvorschläge
Produktvorschläge
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation
Dokumentation
Zertifikate
Aktionen
Aktionen