missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EXT2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 11348-1-AP-150UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
This gene encodes one of two glycosyltransferases involved in the chain elongation step of heparan sulfate biosynthesis. Mutations in this gene cause the type II form of multiple exostoses. Alternatively spliced transcript variants encoding different isoforms have been noted for this gene.Spezifikation
EXT2 | |
Polyclonal | |
Unconjugated | |
EXT2 | |
exostoses (multiple) 2, Exostosin 2, EXT2, Multiple exostoses protein 2, SOTV | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
14043, 2132, 311215 | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Western Blot | |
0.14 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P70428, Q93063 | |
EXT2 | |
EXT2 Fusion Protein Ag1748 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Liquid |