missing translation for 'onlineSavingsMsg'
Learn More

exo-Granisetron, TRC

Product Code. 30004323
Change view
Click to view available options
Menge:
100 mg
Unit Size:
100 Milligramm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Menge Unit Size
30004323 100 mg 100 Milligramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30004323 Supplier Toronto Research Chemicals Supplier No. TRCG780010100MG
 Eingeschränkter Artikel
Verfügbar ab: 30-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Hochreine organische Moleküle und analytische Standards, strategisch weltweit bereitgestellt, um Innovation und kommerziellen Erfolg zu ermöglichen.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1364914-39-5
Summenformel C18H24N4O
Molekulargewicht (g/mol) 312.41
Synonym exo-N-(9-Methyl-9-azabicyclo[3.3.1]non-3-yl)-1-methyl-1H-indazole-3-carboxamide;
SMILES CN1[C@@H]2CCC[C@H]1C[C@@H](C2)NC(=O)c3nn(C)c4ccccc34

Specifications

Chemischer Name oder Material exo-Granisetron (Granisetron Impurity F)
Hinweise zur Reinheitsqualität HPLC
Menge 100 mg
InChI-Formel InChI=1S/C18H24N4O/c1-21-13-6-5-7-14(21)11-12(10-13)19-18(23)17-15-8-3-4-9-16(15)22(2)20-17/h3-4,8-9,12-14H,5-7,10-11H2,1-2H3,(H,19,23)/t12?,13-,14+
Reinheit (%) >95%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.