Learn More
Piperidin-4-carbonsäureethylester, 98%, Thermo Scientific Chemicals
Beschreibung
It is used as a reactant for synthesis of SMN protein modulators, β-aryl and β-amino-substituted aliphatic esters by rhodium catalyzed tandem double bond migration/conjugate addition, nitroethylenediamines by nucleophilic ring opening of nitroimidazolidinone. It is also involved in the reactions of RhoA inhibitors for cardiovascular disease therapy and saccharin derived Mannich bases as antimicrobials and antioxidants. It is employed as a reactant for one-pot reductive amination and Suzuki-Miyaura cross coupling of formyl aryl and heteroaryl MIDA boronates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Vollständig mit Wasser mischbar.
Hinweise
Bei Raumtemperatur lagern. Nicht kompatibel mit starken Oxidationsmitteln.
Spezifikation
Chemischer Name oder Material | Ethyl isonipecotate |
Dichte | 1.021 |
Siedepunkt | 203°C to 205°C |
Flammpunkt | 80°C (176°F) |
Brechungsindex | 1.46 |
Menge | 25 g |
Beilstein | 118419 |
Löslichkeitsinformationen | Fully miscible with water. |
Formelmasse | 157.21 |
Reinheit (%) | 98% |
RUO – Research Use Only