missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4-chlor-6-fluorchinolin-3-carboxylat, 95 %, Thermo Scientific™

Catalog Number 11335579
Click to view available options
Menge:
250 mg
1 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
77779-49-8
C12H9ClFNO2
253.657
MFCD00173350
PPHSOQWURBALSQ-UHFFFAOYSA-N
ethyl4-chloro-6-fluoroquinoline-3-carboxylate, 4-chloro-6-fluoroquinoline-3-carboxylic acid ethyl ester, acmc-1bfwa, ethyl 4-chloro-6-fluoroquinolin-3-carboxylate, ethyl 4-chloro-6-fluoro-3-quinolinecarboxylate, ethyl 6-chloro-4-fluoroquinoline-3-carboxylate, ethyl 4-chloro-6-fluoro-quinoline-3-carboxylate, 4chloro-6-fluoro-quinoline-3-carboxylic acid ethyl ester, 3-quinolinecarboxylicacid, 4-chloro-6-fluoro-, ethyl ester, 4-chloro-6-fluoro-quinoline-3-carboxylic acid ethyl ester
1479088
Ethyl 4-Chlor-6-Fluorchinolin-3-Carboxylat
CCOC(=O)C1=CN=C2C=CC(=CC2=C1Cl)F
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
77779-49-8
C12H9ClFNO2
253.657
MFCD00173350
PPHSOQWURBALSQ-UHFFFAOYSA-N
ethyl4-chloro-6-fluoroquinoline-3-carboxylate, 4-chloro-6-fluoroquinoline-3-carboxylic acid ethyl ester, acmc-1bfwa, ethyl 4-chloro-6-fluoroquinolin-3-carboxylate, ethyl 4-chloro-6-fluoro-3-quinolinecarboxylate, ethyl 6-chloro-4-fluoroquinoline-3-carboxylate, ethyl 4-chloro-6-fluoro-quinoline-3-carboxylate, 4chloro-6-fluoro-quinoline-3-carboxylic acid ethyl ester, 3-quinolinecarboxylicacid, 4-chloro-6-fluoro-, ethyl ester, 4-chloro-6-fluoro-quinoline-3-carboxylic acid ethyl ester
1479088
Ethyl 4-Chlor-6-Fluorchinolin-3-Carboxylat
CCOC(=O)C1=CN=C2C=CC(=CC2=C1Cl)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material Ethyl 4-chloro-6-fluoroquinoline-3-carboxylate
Menge 250 mg
Formelmasse 253.66
Reinheit (%) 95%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'