missing translation for 'onlineSavingsMsg'
Learn More

Ethyl-3-(2-furyl)-1H-pyrazol-5-carboxylat, 98 %, Thermo Scientific Chemicals

Product Code. 11496430
Change view
Click to view available options
Menge:
5 g
25 g
Unit Size:
25 Gramm
5 Gramm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Menge Unit Size
11496430 25 g 25 Gramm
11415500 5 g 5 Gramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11496430 Supplier Thermo Scientific Alfa Aesar Supplier No. H25791.14

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 33545-44-7 | Molecular Formula: C10H10N2O3 | Molecular Weight: 206.20 g/mol

Dieses Markenprodukt von Thermo Scientific Chemicals war ursprünglich Teil des Produktportfolios von Alfa Aesar. Einige Dokumente und Etiketteninformationen enthalten möglicherweise Angaben zur bisherigen Marke. Die ursprüngliche Produkt- oder Artikelnummer bei Alfa Aesar oder die SKU-Referenz wurde im Rahmen des Übergangs der Marke zu Thermo Scientific Chemicals nicht geändert.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 33545-44-7
Summenformel C10H10N2O3
Molekulargewicht (g/mol) 206.20
MDL-Nummer MFCD09953454,MFCD06797478
InChI-Schlüssel GSVBXAZEQDJUOY-UHFFFAOYSA-N
Synonym ethyl 5-2-furyl-1h-pyrazole-3-carboxylate, ethyl 5-furan-2-yl-1h-pyrazole-3-carboxylate, ethyl 3-furan-2-yl-1h-pyrazole-5-carboxylate, ethyl 5-fur-2-yl-1h-pyrazole-3-carboxylate, ethyl 5-2-furyl pyrazole-3-carboxylate, ethyl 5-furan-2-yl-2h-pyrazole-3-carboxylate, aronis23991, ethyl 3-2-furyl pyrazole-5-carboxylate, 1h-pyrazole-3-carboxylicacid, 5-2-furanyl-, ethyl ester
PubChem CID 7127811
IUPAC-Name Ethyl 3-(Furan-2-yl)-1H-Pyrazol-5-Carboxylat
SMILES CCOC(=O)C1=CC(=NN1)C1=CC=CO1

Specifications

Chemischer Name oder Material Ethyl 3-(2-furyl)-1H-pyrazole-5-carboxylate
Schmelzpunkt 135°C to 137°C
Menge 25 g
Formelmasse 206.2
Reinheit (%) 98%

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.