missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ETFA Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 12262-1-AP-20UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
ETFA participates in catalyzing the initial step of the mitochondrial fatty acid beta-oxidation. It shuttles electrons between primary flavoprotein dehydrogenases and the membrane-bound electron transfer flavoprotein ubiquinone oxidoreductase. Defects in electron-transfer-flavoprotein have been implicated in type II glutaricaciduria in which multiple acyl-CoA dehydrogenase deficiencies result in large excretion of glutaric, lactic, ethylmalonic, butyric, isobutyric, 2-methyl-butyric, and isovaleric acids. Two transcript variants encoding different isoforms have been found for this gene.Spezifikation
ETFA | |
Polyclonal | |
Unconjugated | |
ETFA | |
Alpha ETF, EMA, ETFA, GA2, MADD | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
110842, 2108, 300726 | |
Antibody | |
IgG |
Flow Cytometry, Western Blot, Immunofluorescence, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
0.19 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P13803, P13804, Q99LC5 | |
ETFA | |
ETFA Fusion Protein Ag2931 | |
20 μL | |
Primary | |
Rat, Mouse, Human | |
Liquid |