missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ETEA Polyclonal Antibody
Rabbit Polyclonal Antibody
Marke: Invitrogen™ PA521887
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Recommended positive controls: H1299, HeLa. Predicted reactivity: Mouse (100%), Rat (100%), Rhesus Monkey (100%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The protein encoded by this gene is highly expressed in peripheral blood of patients with atopic dermatitis (AD), compared to normal individuals. It may play a role in regulating the resistance to apoptosis that is observed in T cells and eosinophils of AD patients.
Spezifikation
| ETEA | |
| Polyclonal | |
| Unconjugated | |
| FAF2 | |
| 2210404d11rik; AI462440; ETEA; expressed in T-cells and eosinophils in atopic dermatitis; FAF2; Fas associated factor family member 2; FAS-associated factor 2; Kiaa0887; mKIAA0887; Protein ETEA; protein expressed in T-cells and eosinophils in atopic dermatitis; RGD1306577; UBX domain containing 8; UBX domain protein 3B; UBX domain-containing protein 3B; UBX domain-containing protein 8; UBX8; UBXD8; UBXN3B; Unknown (protein for MGC:129072) | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 23197, 76577 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.57 mg/mL | |
| 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
| Q3TDN2, Q96CS3 | |
| FAF2 | |
| Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human ETEA. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur