Learn More
Esmolol Hydrochloride, TRC
Artikelnummer. 17951355
Change view
Click to view available options
Menge:
10 mg
25 mg
Packungsgröße:
10 Milligramm
25 Milligramm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge unitSize
17951355 25 mg 25 Milligramm
17910865 10 mg 10 Milligramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 17951355 Lieferant Toronto Research Chemicals Lieferanten-Nr. TRCE66800025MG
om dit product te kopen Registreer vandaag om een webaccount aan te maken
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemische Identifikatoren
CAS 81161-17-3
Summenformel C16 H25 N O4 . Cl H
Molekulargewicht (g/mol) 331.83
Synonym Benzenepropanoic acid, 4-[2-hydroxy-3-[(1-methylethyl)amino]propoxy]-, methyl ester, hydrochloride (1:1), Benzenepropanoic acid, 4-[2-hydroxy-3-[(1-methylethyl)amino]propoxy]-, methyl ester, hydrochloride (9CI), ASL 8052, Brevibloc, Clol, Esmolol hydrochloride
IUPAC-Name methyl 3-[4-[2-hydroxy-3-(propan-2-ylamino)propoxy]phenyl]propanoate;hydrochloride
SMILES Cl.COC(=O)CCc1ccc(OCC(O)CNC(C)C)cc1
Spezifikation
Chemischer Name oder Material Esmolol Hydrochloride
Hinweise zur Reinheitsqualität HPLC
Menge 25 mg
InChI-Formel InChI=1S/C16H25NO4.ClH/c1-12(2)17-10-14(18)11-21-15-7-4-13(5-8-15)6-9-16(19)20-3;/h4-5,7-8,12,14,17-18H,6,9-11H2,1-3H3;1H
Formelmasse 331.155
Reinheit (%) >95%