Learn More
ERG Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke:  Proteintech 14356-1-AP-150UL
 Lernen Sie weitere Varianten dieses
Artikelnummer. 16822964
  • 385.92 CHF / 150 Mikroliter
om dit product te kopen Registreer vandaag om een webaccount aan te maken
Beschreibung
Beschreibung
Transcriptional regulator. May participate in transcriptional regulation through the recruitment of SETDB1 histone methyltransferase and subsequent modification of local chromatin structure.
Spezifikation
Spezifikation
ERG
Polyclonal
Unconjugated
ERG
ERG, erg 3, p55, Transcriptional regulator ERG, Transforming protein ERG
Rabbit
Antigen Affinity Chromatography
RUO
13876, 170909, 2078
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
0.27 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P11308, P81270
Erg
ERG Fusion Protein Ag5708
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Produktvorschläge
Produktvorschläge
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation
Dokumentation
Zertifikate
Aktionen
Aktionen