Learn More
Epsin 3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA563142
Additional Details : Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: SADPWDIPGF RPNTEASGSS WGPSADPWSP IPSGTVLSRS QPWDLTPMLS SSEPWGRTPV LPAGPPTTDP WALNSPHHKL PST Highest antigen sequence identity to the following orthologs: Mouse - 83%, Rat - 84%.
EPN3 belongs to the epsin family.Spezifikation
Epsin 3 | |
Polyclonal | |
Unconjugated | |
EPN3 | |
2310022G12Rik; Epn3; EPS-15-interacting protein 3; epsin 3; epsin-3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55040 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9H201 | |
EPN3 | |
Recombinant protein corresponding to Human Epsin 3 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only