missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EPS15R Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA550339
Additional Details : Gewicht : 0.01000kg
Beschreibung
The antibody detects endogenous levels of total EPS15R protein.
EPS15R Polyclonal antibody specifically detects EPS15R in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Spezifikation
EPS15R | |
Polyclonal | |
Unconjugated | |
EPS15L1 | |
9830147J04Rik; AI593686; epidermal growth factor receptor pathway substrate 15 like 1; epidermal growth factor receptor pathway substrate 15-like 1; epidermal growth factor receptor pathway substrate 15-related sequence; Epidermal growth factor receptor substrate 15-like 1; epidermal growth factor receptor substrate EPS15R; Eps15l1; EPS15R; eps15-related protein; Eps15-rs | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
58513 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1.4 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Q9UBC2 | |
EPS15L1 | |
Fusion protein corresponding to residues near the C terminal of human epidermal growth factor receptor pathway substrate 15-like 1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |