missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPINLW1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA550656
Additional Details : Gewicht : 0.01000kg
Beschreibung
The antibody detects endogenous levels of total EPPIN protein.
SPINLW1 Polyclonal antibody specifically detects SPINLW1 in Human, Mouse samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Spezifikation
SPINLW1 | |
Polyclonal | |
Unconjugated | |
EPPIN | |
1700024E17Rik; cancer/testis antigen 71; cancer/testis antigen 72; CT71; CT72; dJ461P17.2; epididymal peptidase inhibitor; epididymal protease inhibitor; EPPIN; EPPIN1; EPPIN2; EPPIN3; protease inhibitor WAP7; serine peptidase inhibitor-like, with Kunitz and WAP domains 1 (eppin); serine protease inhibitor-like with Kunitz and WAP domains 1; serine protease inhibitor-like, with Kunitz and WAP domains 1 (eppin); Spinlw1; WAP four-disulfide core domain protein 7; WAP four-disulfide core domain protein 8; WAP7; WFDC7; Wfdc8 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
57119, 75526 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
2.4 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
O95925, Q9DA01 | |
EPPIN | |
Synthetic peptide corresponding to a region derived from internal residues of human epididymal peptidase inhibitor | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |