Learn More
Invitrogen™ Epiregulin (EREG) Mouse ELISA Kit
Sandwich ELISA Kit
Marke: Invitrogen™ EMEREGX5
Additional Details : Gewicht : 0.33800kg
Beschreibung
The Mouse Epiregulin (Ms EREG) ELISA quantitates Ms Epiregulin in mouse serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Ms Epiregulin. Principle of the method The Mouse Epiregulin solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
EREG ist ein Familienmitglied der epidermalen Wachstumsfaktoren. EREG kann als Ligand von EGFR (epidermaler Wachstumsfaktor-Rezeptor) sowie als Ligand der meisten ERBB-Familienmitglieder (v-erb-b2 Onkogen-Homolog) von Tyrosinkinase-Rezeptoren fungieren.Spezifikation
Q61521 | |
10 pg/mL | |
Festphasen-Sandwich-ELISA | |
ELISA-Kit | |
Plasma, Serum, Überstand | |
5 x 96 Assays | |
ELISA, Protein-Assays, Proteinbiologie, Krebsbiologie, Immunologie, Neurowissenschaften, Neurobiologie, Proteinnachweis, Proteinanalyse | |
13874 | |
<12% | |
Vorbeschichtete 96-Well-Platten, Biotinylierter Nachweisantikörper, Assay-Verdünnungskonzentrat, Standard, Dichtungsfolien für Mikrotiterplatten, Chromogen, SAV-HRP, Waschpuffer, Stopplösung | |
5 x 96 Tests | |
Angiogenese | |
2°bis 8 °C | |
Maus | |
4 h 45 min |
8,23 pg/mL bis 6000 pg/mL | |
10 pg/mL bis 6000 pg/mL | |
Biotin | |
Wachstumsfaktoren und Rezeptoren | |
Dieses ELISA-Kit zeigt keine Kreuzreaktivität mit den folgenden getesteten Zytokinen: Maus CD30, L CD30, T CD40, CRG-2, CTACK, CXCL16, Eotaxin, Eotaxin-2, Fas Ligand, Fraktalkin, GCSF, GM-CFS, IFN-gamma, IGFBP-3, IGFBP-5, IGFBP-6, IL-1 alpha, IL-1 beta, IL-2, IL-3, IL-3 Rb, IL-4, IL-5, IL-9, IL-10, IL-12 p40/p70, IL-12 p70, IL-13, IL-17, KC, Leptin R, LEPTIN (OB), LIX, L-Selektin, Lymphotactin, MCP-1, MCP-5, M-CSF, MIG, MIP-1 alpha, MIP-1 gamma, MIP-2, MIP-3 beta, MIP-3 alpha, PF-4, P-Selektin, RANTES, SCF, SDF-1 alpha, TARC, TCA-3, TECK, TIMP-1, TNF-alpha, TNF RI, TNF RII, TPO, VCAM-1, VEGF. | |
Maus | |
Kolorimetrisches Mikrotiterplatten-Lesegerät | |
Ereg | |
<10% | |
HRP | |
RUO | |
Plasma, 50 μl; Serum, 50 μl; Überstand, 100 μl | |
EPR | |
1 h 20 min |