Learn More
EphA4 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA529338
Additional Details : Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: HeLa, 293T, mouse brain, mouse hippocampus, rat brain, A2058, DDDDK-tagged EPHA4-transfected 293T. Predicted reactivity: Mouse (100%), Rat (99%), Xenopus laevis (92%), Pig (99%), Chicken (96%), Rhesus Monkey (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
EPHA4 belongs to the ephrin receptor subfamily of the protein tyrosine kinase family. EPH and EPH-related receptors have been implicated in mediated developmental events, particularly in the nervous system. Receptors in the EPH subfamily typically have a single kinase domain and an extracellular region containing a Cys-rich domain and 2 fibronectin type III repeats. The ephrin receptors are divided into 2 groups based on the similarity of their extracellular domain sequences and their affinities for binding ephrin-A and ephrin-B ligands. EphA4 encodes a protein that binds ephrin-A ligands.Spezifikation
EphA4 | |
Polyclonal | |
Unconjugated | |
EPHA4 | |
2900005C20Rik; AI385584; Cek8; Cek8 protein; EK8; epa4; EPH receptor A4; Epha4; EPH-like kinase 8; Ephrin type-A receptor 4; Hek8; rb; receptor protein-tyrosine kinase HEK8; RGD1560587; Sek; Sek1; Tyro1; TYRO1 protein tyrosine kinase; tyrosine-protein kinase receptor MPK-3; Tyrosine-protein kinase receptor SEK; tyrosine-protein kinase receptor SEK-1; Tyrosine-protein kinase TYRO1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
13838, 2043, 316539 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.97 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
P54764, Q03137 | |
EPHA4 | |
Recombinant fragment corresponding to a region within amino acids 430 and 766 of Human Eph receptor A4 | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |