missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EPHA1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 18698-1-AP-150UL
Additional Details : Gewicht : 0.00010kg
Descrizione
This gene belongs to the ephrin receptor subfamily of the protein-tyrosine kinase family. EPH and EPH-related receptors have been implicated in mediating developmental events, particularly in the nervous system. Receptors in the EPH subfamily typically have a single kinase domain and an extracellular region containing a Cys-rich domain and 2 fibronectin type III repeats. The ephrin receptors are divided into 2 groups based on the similarity of their extracellular domain sequences and their affinities for binding ephrin-A and ephrin-B ligands. This gene is expressed in some human cancer cell lines and has been implicated in carcinogenesis.Specifica
EPHA1 | |
Polyclonal | |
Unconjugated | |
EPHA1 | |
EPH, EPH receptor A1, EPHA1, Ephrin type A receptor 1, EPHT, EPHT1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
13835, 2041 | |
-20°C | |
Liquid |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation | |
0.17 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P21709, Q60750 | |
EPHA1 | |
Peptide | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |