Learn More
EPCR Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA532217
Additional Details : Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: 293T, JC, BCL-1. Predicted reactivity: Dog (80%), Pig (82%), Bovine (82%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
EPCR Polyclonal antibody specifically detects EPCR in Human samples. It is validated for Western Blot.Spezifikation
EPCR | |
Polyclonal | |
Unconjugated | |
Procr | |
Activated protein C receptor; AI325044; APC receptor; bA42O4.2; CCCA; CCD41; CD antigen CD201; CD201; CD201 antigen; cell cycle, centrosome-associated protein; centrocyclin; Centrosomal protein CCD41; endothelial cell protein C receptor; Endothelial protein C receptor; Epcr; PROCR; protein C receptor; protein C receptor, endothelial | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10544 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1.34 mg/mL | |
PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
Q9UNN8 | |
Procr | |
Recombinant fragment corresponding to a region within amino acids 121 and 238 of Human EPCR | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |