Learn More
EPB41 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA565015
Additional Details : Gewicht : 0.01000kg
Beschreibung
Immunogen sequence: ESVPEPRPSEW DKRLSTHSPF RTLNINGQIP TGEGPPLVKT QTVTISDNAN AVKSEIPTKD VPIVHTETKT Highest antigen sequence identity to the following orthologs - mouse 96%, rat 96%.
EPB41 Polyclonal antibody specifically detects EPB41 in Human samples. It is validated for Immunocytochemistry.Spezifikation
EPB41 | |
Polyclonal | |
Unconjugated | |
EPB41 | |
4.1R; AI415518; Band 4.1; D4Ertd442e; E41P; EL1; elliptocytosis 1, RH-linked; Elp1; Elp-1; EPB4.1; EPB41; erythrocyte membrane protein band 4.1; erythrocyte membrane protein band 4.1 (elliptocytosis 1, RH-linked); erythrocyte protein band 4.1; erythrocyte skeletal protein; erythrocyte surface protein band 4.1; HE; Kiaa4056; mKIAA4056; P4.1; protein 4.1; Protein 4.1R; RGD1564762 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
2035 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P11171 | |
EPB41 | |
Recombinant Human EPB41 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
For Research Use Only