Learn More
EPB41 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA565025
Additional Details : Gewicht : 0.01000kg
Beschreibung
Immunogen sequence: EKSLVTEAENS QHQQKEEGEE AINSGQQEPQ QEESCQTAAE GDNWCEQKLK ASNGDTPTHE DLTKNKERTS ESRGLSRLFS SFLKRPKSQV SE Highest antigen sequence identity to the following orthologs - mouse 83%, rat 84%.
Elliptocytosis is a hematologic disorder characterized by elliptically shaped erythrocytes and a variable degree of hemolytic anemia. Inherited as an autosomal dominant, elliptocytosis results from mutation in any one of several genes encoding proteins of the red cell membrane skeleton. The form discussed here is the one found in the 1950s to be linked to Rh blood group and more recently shown to be caused by a defect in protein 4. 1. 'Rh-unlinked' forms of elliptocytosis are caused by mutation in the alpha-spectrin gene, the beta-spectrin gene, or the band 3 gene.Spezifikation
EPB41 | |
Polyclonal | |
Unconjugated | |
EPB41 | |
4.1R; AI415518; Band 4.1; D4Ertd442e; E41P; EL1; elliptocytosis 1, RH-linked; Elp1; Elp-1; EPB4.1; EPB41; erythrocyte membrane protein band 4.1; erythrocyte membrane protein band 4.1 (elliptocytosis 1, RH-linked); erythrocyte protein band 4.1; erythrocyte skeletal protein; erythrocyte surface protein band 4.1; HE; Kiaa4056; mKIAA4056; P4.1; protein 4.1; Protein 4.1R; RGD1564762 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
2035 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Western Blot | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P11171 | |
EPB41 | |
Recombinant Human EPB41 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
For Research Use Only