missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Endothelin 1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 12191-1-AP-20UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
The protein encoded by this gene is proteolytically processed to release a secreted peptide termed endothelin 1. This peptide is a potent vasoconstrictor and is produced by vascular endothelial cells. Endothelin 1 also can affect the central nervous system. Two transcript variants encoding different isoforms have been found for this gene.Spezifikation
Endothelin 1 | |
Polyclonal | |
Unconjugated | |
EDN1 | |
EDN1, endothelin 1, ET 1 Big endothelin 1, ET1, HDLCQ7, PPET1, Preproendothelin 1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
1906 | |
Antibody | |
IgG |
Western Blot | |
0.37 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P05305 | |
EDN1 | |
Endothelin 1 Fusion Protein Ag2847 | |
20 μL | |
Primary | |
Human | |
Liquid |