missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ EMP Polyclonal Antibody
Sheep Polyclonal Antibody
Marke: Invitrogen™ PA547914
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Reconstitute in sterile PBS to a final concentration of 0.2 mg/mL.
This gene product mediates the attachment of erythroblasts to macrophages. This attachment promotes terminal maturation and enucleation of erythroblasts, presumably by suppressing apoptosis. This protein is an integral membrane protein with the N-terminus on the extracellular side and the C-terminus on the cytoplasmic side of the cell. Two immunologically related isoforms of erythroblast macrophage protein with apparent molecular weights of 33 kD and 36 kD were detected in macrophage membranes, this gene encodes the larger isoform. Multiple alternatively spliced transcript variants encoding distinct isoforms have been found for this gene, but the biological validity of some variants has not been determined.
Spezifikation
| EMP | |
| Polyclonal | |
| Unconjugated | |
| MAEA | |
| 1110030D19Rik; Cell proliferation-inducing gene 5 protein; E3 ubiquitin-protein transferase MAEA; EMLP; EMP; Erythroblast macrophage protein; GID complex subunit 9, FYV10 homolog; GID9; HLC10; HLC-10; Human lung cancer oncogene 10 protein; lung cancer-related protein 10; macrophage erythroblast attacher; MAEA; P44EMLP; PIG5 | |
| Sheep | |
| Antigen affinity chromatography | |
| RUO | |
| 10296, 298982, 59003 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Western Blot, Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 5% trehalose and No Preservative | |
| Q4VC33, Q5RKJ1, Q7L5Y9 | |
| MAEA | |
| E. coli-derived recombinant human EMP/MAEA Met12-Ser66. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur