Learn More
Euro Protection™ Emerald High-Cut Safety Shoes
Euro Protection™ Emerald High-Cut Safety Shoes are comfortable, breathable, and feature a composite toe cap for protection
Marke:  Euro Protection™ 9EMEH43
Additional Details : Gewicht : 0.01000kg
 Lernen Sie weitere Varianten dieses
Artikelnummer. 12088326
  • 95.69 CHF / 1 Stück
Voraussichtliches Versanddatum: 20-08-2021
Zum Warenkorb hinzufügen
Beschreibung
Beschreibung
  • Breathable upper and lining.
  • Padding for snug fit.
  • Rounded heel for more comfortable driving.
  • Nubuck leather and nylon mesh combination upper.
  • Moisture wicking non-woven lining.
  • Attached, textile and foam insole.
  • Composite toe cap.
  • Non-metallic (composite fabric) puncture resistant sole.
  • Injected polyurethane midsole.
  • Polyurethane outsole.
Spezifikation
Spezifikation
Tan, Yellow
Leather, Nylon
Attached textile and foam insole, non-metallic (composite fabric) puncture-resistant sole, injected polyurethane midsole, polyurethane outsole.
High
692 g per shoe in size 8
Moisture wicking non-woven lining
43
Polyurethane, Composite Fabric
Composite
EN ISO 20345 : 2011