missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ELOF1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 21070-1-AP-150UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
Eukaryotic RNA polymerase II mediates the synthesis of mature and functional messenger RNA. This is a multistep process, called the transcription cycle, that includes five stages: preinitiation, promoter, clearance, elongation and termination. Elongation is thought to be a critical stage for the regulation of gene expression. ELOF1 (Transcription elongation factor 1 homolog) is a 83 amino acid nuclear protein that is implicated in the maintece of proper chromatin structure in regions that are being actively transcribed. The gene encoding ELOF1 maps to human chromosome 19, which consists of over 63 million bases, houses approximately 1,400 genes and is recognized for having the greatest gene density of the human chromosomes. It is the genetic home for a number of immunoglobulin (Ig) superfamily members, including the killer cell and leukocyte Ig-like receptors, a number of ICAMs, the CEACAM and PSG family and Fc receptors (FcRs).Spezifikation
ELOF1 | |
Polyclonal | |
Unconjugated | |
ELOF1 | |
ELF1, ELOF1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
66126, 691193, 84337 | |
-20°C | |
Liquid |
Western Blot | |
0.29 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P60002, P60003 | |
Elof1 | |
ELOF1 Fusion Protein Ag15213 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |