missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PL BioScience ELAREM™ Perform-FD PLUS, Fibrinogen-depleted Human Platelet Lysate, No Heparin Added or Required
Click to view available options
Menge:
50 ml
100 ml
500 ml
Unit Size:
100 Milliliter
50 Milliliter
500 Milliliter
Description
- Clot-free and Fibrinogen-depleted HPL: ELAREM™ Perform-FD PLUS is a fibrinogen-depleted, ready-to-use Human Platelet Lysate that is widely applicable.
- No Heparin Added or Required: Due to the unique fibrinogen-depletion technique, ELAREM™ Perform-FD PLUS is clot-free and heparin-free. The product neither requires the addition of anticoagulants (e.g. heparin) in cell culture nor during manufacturing.
- High Performance Lab Process: ELAREM™ Perform-FD PLUS efficiently and safely supports in vitro expansion of various primary cells and cell lines.
- EU Origin for Long Traceability: ELAREM™ Perform-FD PLUS is manufactured from platelet units obtained from healthy blood donors at licensed blood centers in the EU. ELAREM™ Perform-FD PLUS is derived from EU origin platelet units which ensures long raw material traceability periods.
- Intended Use: ELAREM™ Perform-FD PLUS is for in vitro and manufacturing use only. The product is not intended for direct therapeutic use. For safety precautions, please see appropriate Safety Data Sheet (SDS).
Specifications
Specifications
| Farbe | Yellow |
| Kulturtyp | Adherent Cell Culture, Suspension Cell Culture |
| Form | Flüssig |
| Produkttyp | Zellkultur-Supplement |
| Sterilität | Steril |
| Sterilization Method | Steril gefiltert |
| Zusammensetzung | Liquid, Human Platelet Lysate, Fibrinogen-depleted |
| Reinheits- oder Qualitätsgrad | Forschungsqualität |
| Menge | 50 ml |
| Versandbedingung | Dry Ice |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction