Learn More
Invitrogen™ Total Antioxidant Capacity (T-AOC) Colorimetric Assay Kit (ABTS, enzyme method)
Beschreibung
The Total Antioxidant Capacity (T-AOC) Colorimetric Assay Kit (ABTS, enzyme method) is a highly reliable and sensitive assay designed for the quantitative measurement of T-AOC in serum, plasma, cells, and other biological sample types. T-AOC refers to the overall ability of a biological sample to counteract oxidative stress by neutralizing reactive oxygen species (ROS) and other harmful free radicals. It represents the cumulative effect of both enzymatic and non-enzymatic antioxidants present in the sample.
The assay uses ABTS, which is oxidized to ABTS+ in solution. ABTS+ is measured at 414 or 734 nm and is proportional to the T-AOC in the sample.
Key features
• Simple and fast protocol—user-friendly, requiring minimal hands-on time and providing rapid results within a short incubation period
• Broad range of kits—across key analytes and targets within the most important metabolic pathways
• Consistent—strict quality control criteria to ensure reproducible and reliable results
Spezifikation
Spezifikation
| Assay-Bereich | 0.047–1.50 mmol/L |
| Assay-Empfindlichkeit: | 0.047 mmol/L |
| Inhalt und Lagerung | Reagent 5: –20°C Other reagents: 2–8°C |
| Nachweisverfahren | Kolorimetrisch |
| Probentyp | Plasma, Speichel, Serum, Tissue Homogenates, Urin, Whole Cells |
| Zur Verwendung mit (Geräte) | Mikrotiterplatten-Lesegerät |
| Inkubationszeit | 6 min. |
| Interassay-VK | 4.1 |
| Intraassay-VK | 2.2 |
| Menge | 96 Tests |
| Mehr anzeigen |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.