missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EIF3M Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 11423-1-AP-150UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
This gene encodes a protein that is part of the eurkaryotic translation initiation factor 3 complete (eIF-3) required for protein synthesis. Elevated levels of the encoded protein are present in cancer cell lines. Inactivation of the encoded protein has been shown to interfere with translation of herpes virus mRNAs by preventing the association of mRNAs with the ribosomes. A pseudogene of this gene is located on the X chromosome.Spezifikation
EIF3M | |
Polyclonal | |
Unconjugated | |
EIF3M | |
B5, EIF3M, Fetal lung protein B5, GA17, hfl B5, HFLB5, PCID1, PNAS 125 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10480, 295975, 98221 | |
Antibody | |
IgG |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q7L2H7, Q99JX4 | |
EIF3M | |
EIF3M Fusion Protein Ag1994 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Liquid |