Learn More
EIF2S3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke:  Proteintech 11162-1-AP-20UL
 Lernen Sie weitere Varianten dieses
Artikelnummer. 16808053
  • 136.48 CHF / 20 Mikroliter
om dit product te kopen Registreer vandaag om een webaccount aan te maken
Beschreibung
Beschreibung
The initiation of protein synthesis in eukaryotic cells is regulated by interactions between protein initiation factors and RNA molecules. The eukaryotic initiation complex is composed of three subunits, designated eIF2alpha, eIF2beta and eIF2gamma (eukaryotic translation initiation factor 2 alpha, beta and gamma, respectively), all of which work in concert to form a ternary complex with GTP and tRNA in the early stages of protein synthesis. eIF2S3 (eukaryotic translation initiation factor 2, subunit 3), also known as eIF2G, is a 472 amino acid protein that belongs to the gamma subfamily of GTP-binding elongation factor proteins. Existing as a heterotrimer composed of an alpha, beta and gamma chain, eIF2S3 functions to bind ribosomal subunits and catalyze the subsequent formation of preinitiationcomplexes necessary for protein synthesis.
Spezifikation
Spezifikation
EIF2S3
Polyclonal
Unconjugated
EIF2S3
eIF 2 gamma X, eIF 2gA, eIF 2gX, EIF2, EIF2G, EIF2gamma, EIF2S3
Rabbit
Antigen Affinity Chromatography
RUO
1968
-20°C
Liquid
Immunocytochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunofluorescence, Western Blot
0.25 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P41091
EIF2S3
EIF2S3 Fusion Protein Ag1650
20 μL
Primary
Human, Mouse
Antibody
IgG
Produktvorschläge
Produktvorschläge
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation
Dokumentation
Zertifikate
Aktionen
Aktionen