missing translation for 'onlineSavingsMsg'
Learn More

EHMT2 Mouse anti-Human, Mouse, Clone: 1A12E3, Proteintech

Mouse Monoclonal Antibody

Marke:  Proteintech 66689-1-IG-150UL

Additional Details : Gewicht : 0.00010kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 16838805

  • 381.34 CHF / 150 Mikroliter
Verfügbar ab: 16-04-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

This gene encodes an enzyme, consisting of two identical proteins, which catalyzes the isomerization of glyceraldehydes 3-phosphate and dihydroxy-acetone phosphate in glycolysis and gluconeogenesis. Mutations in this gene are associated with triosephosphate isomerase deficiency. Pseudogenes have been identified on chromosomes 1, 4, 6 and 7. Alternative splicing results in multiple transcript variants.
Spezifikation

Spezifikation

EHMT2
Monoclonal
1 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q96KQ7, Q9Z148
EHMT2
EHMT2 Fusion Protein Ag27477
150 μL
Primary
Human, Mouse
Antibody
IgG2a
Western Blot
1A12E3
Unconjugated
EHMT2
BAT8, C6orf30, DKFZp686H08213, EHMT2, FLJ35547, G9A, H3 K9 HMTase 3, HLA B associated transcript 8, KMT1C, Lysine N methyltransferase 1C, NG36, NG36/G9a, Protein G9a
Mouse
Protein A
RUO
10919, 110147
-20°C
Liquid
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote