missing translation for 'onlineSavingsMsg'
Learn More

ACROBiosystems Ebolavirus (subtype Bundibugyo, strain Uganda 2007) Envelope Glycoprotein 1 (GP1) Protein, His Tag

Marke:  ACROBiosystems BEP-V5221-100UG

Additional Details : Gewicht : 0.00001kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 15826047

  • 325.15 CHF / 100 Mikrogramm
Verfügbar ab: 11-04-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

EBOV encodes seven structural proteins: nucleoprotein (NP), polymerase cofactor (VP35), (VP40), GP, transcription activator (VP30), VP24, and RNA polymerase (L). GP protein contains 160-kDa envelope-attached glycoprotein (GP) and a 110 kDa secreted glycoprotein (sGP). GP is a class I fusion protein which assembles as trimers on viral surface and plays an important role in virus entry and attachment. Mature GP is a disulfide-linked heterodimer formed by two subunits, GP1 and GP2, which are generated from the proteolytical process of GP precursor (pre-GP) by cellular furin during virus assembly . GP1 is responsible for binding to the receptor(s) on target cells. Interacts with CD209/DC-SIGN and CLEC4M/DC-SIGNR which act as cofactors for virus entry into the host cell. GP2 acts as a class I viral fusion protein. GP1,2 mediates endothelial cell activation and decreases endothelial barrier function. sGP seems to possess an anti-inflammatory activity as it can reverse the barrier-decreasing effects of TNF alpha.

This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 31.8 kDa. The protein migrates as 40-60 kDa under reducing (R) condition (SDS-PAGE).
Spezifikation

Spezifikation

31.8 kDa
Ebolavirus (subtype Bundibugyo, strain Uganda 2007) GP1, His Tag (BEP-V5221) is expressed from human 293 cells (HEK293). It contains AA Ile 33 - Gln 304 (Accession # ACI28624).
-20°C
Glycoprotein / GP (virus)
Ebolavirus
PBS, pH7.4
HEK293
Viral Proteins
100 μg
Ile 33 - Gln 304
GP,Envelope glycoprotein,GP(1,2)
Unconjugated
His Tag
Less than 1.0 EU / μg by the LAL method.
Powder
>90% as determined by SDS-PAGE
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote