Learn More
ACROBiosystems Ebolavirus EBOV (subtype Zaire, strain Kikwit-95) Envelope Glycoprotein 1 (GP1) Protein, His Tag
327.86 CHF - 2105.87 CHF
Spezifikation
Zugriffsnummer | AAQ55048.1 |
---|---|
Molekulargewicht | 52.0 kDa |
Quelle | Ebolavirus EBOV (subtype Zaire, strain Kikwit-95) GP1, His Tag (ZE5-V5220) is expressed from human 293 cells (HEK293). It contains AA Ile 33 - Arg 501 (Accession # AAQ55048.1). |
Immunogen | Ile 33 - Arg 501 |
Lagerungsbedingungen | -20°C |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15827957
|
ACROBiosystems
ZE5-V5220-1MG |
1 mg |
2105.87 CHF
1 Milligramm |
Verfügbar ab: 13-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15817957
|
ACROBiosystems
ZE5-V5220-100UG |
100 μg |
327.86 CHF
100 Mikrogramm |
Verfügbar ab: 13-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Beschreibung
EBOV encodes seven structural proteins: nucleoprotein (NP), polymerase cofactor (VP35), (VP40), GP, transcription activator (VP30), VP24, and RNA polymerase (L). GP protein contains 160-kDa envelope-attached glycoprotein (GP) and a 110 kDa secreted glycoprotein (sGP). GP is a class I fusion protein which assembles as trimers on viral surface and plays an important role in virus entry and attachment. Mature GP is a disulfide-linked heterodimer formed by two subunits, GP1 and GP2, which are generated from the proteolytical process of GP precursor (pre-GP) by cellular furin during virus assembly . GP1 is responsible for binding to the receptor(s) on target cells. Interacts with CD209/DC-SIGN and CLEC4M/DC-SIGNR which act as cofactors for virus entry into the host cell. GP2 acts as a class I viral fusion protein. GP1,2 mediates endothelial cell activation and decreases endothelial barrier function. sGP seems to possess an anti-inflammatory activity as it can reverse the barrier-decreasing effects of TNF alpha.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 52.0 kDa. The protein migrates as 95-120 kDa under reducing (R) condition (SDS-PAGE).Spezifikation
AAQ55048.1 | |
Ebolavirus EBOV (subtype Zaire, strain Kikwit-95) GP1, His Tag (ZE5-V5220) is expressed from human 293 cells (HEK293). It contains AA Ile 33 - Arg 501 (Accession # AAQ55048.1). | |
-20°C | |
Glycoprotein / GP (virus) | |
Ebolavirus | |
PBS, pH7.4 | |
HEK293 | |
Viral Proteins |
52.0 kDa | |
Ile 33 - Arg 501 | |
GP,Envelope glycoprotein,GP(1,2) | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |