Learn More
EBF4 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA5113949
Additional Details : Gewicht : 0.01000kg
Beschreibung
Immunogen sequence: GVPGSPSFLN GSTATSPFAK ERLRPRAAPP KLPTPGLPQS PRRGASRPVF For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Human: 100%; Mouse: 79%
EBF4 belongs to the COE family and contains 1 IPT/TIG domain. EBF4 is a transcriptional factor which recognizes variations of the palindromic sequence 5'-ATTCCCNNGGGAATT-3'. EBF4 belongs to the conserved Olf/EBF family of helix-loop-helix transcription factors, members of which play important roles in neural development and B-cell maturation (Wang et al., 2002 [PubMed 12139918]).[supplied by OMIM].Spezifikation
EBF4 | |
Polyclonal | |
Unconjugated | |
EBF4 | |
COE4; early B cell factor 4; early B-cell factor 3; early B-cell factor 4; early B-cell factor 4-like; EBF family member 4; Ebf3; EBF4; EBF-4; KIAA1442; O/E-4; OE-4; Olf-1; Olf-1/EBF-like 4; RP5-860F19.3; transcription factor COE4 | |
Rabbit | |
Affinity chromatography | |
RUO | |
57593 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9BQW3 | |
EBF4 | |
Synthetic peptide directed towards the C terminal region of human EBF4 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |