missing translation for 'onlineSavingsMsg'
Learn More
Learn More
E-Selectin/CD62E Antibody (RM0034-9J12) - Azide and BSA Free, Novus Biologicals™
Rat Monoclonal Antibody
260.00 CHF - 627.00 CHF
Spezifikation
| Antigen | E-Selectin/CD62E |
|---|---|
| Klon | RM0034-9J12 |
| Konzentration | 0.5 mg/mL |
| Verdünnung | Flow Cytometry 1:100-1:1000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:100-1:400 |
| Anwendungen | Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|
18253844
|
Novus Biologicals
NB110-85473-100UG |
100 μg |
627.00 CHF
100 Mikrogramm |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
|
18282234
|
Novus Biologicals
NB110-85473-0.025MG |
0.025 mg |
260.00 CHF
25 Mikrogramm |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
Beschreibung
E-Selectin/CD62E Monoclonal specifically detects E-Selectin/CD62E in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.Spezifikation
| E-Selectin/CD62E | |
| 0.5 mg/mL | |
| Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rat | |
| A 0.2 um filtered solution in PBS with No Preservative | |
| 6401 | |
| This antibody was produced from a hybridoma (mouse myeloma fused with spleen cells from a rat immunized with cytokine stimulated-mouse endothelial membrane protein. | |
| Primary | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| RM0034-9J12 | |
| Flow Cytometry 1:100-1:1000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:100-1:400 | |
| Monoclonal | |
| Purified | |
| Mouse | |
| CD62 antigen-like family member E, CD62E, CD62E antigen, ELAM, ELAM-1, ELAM1E-selectin, endothelial adhesion molecule 1, Endothelial leukocyte adhesion molecule 1, ESEL, LECAM2, leukocyte endothelial cell adhesion molecule 2, Leukocyte-endothelial cell adhesion molecule 2, selectin E | |
| SELE | |
| IgG2 | |
| Protein G purified | |
| This is specific for mouse E-Selectin |
For Research Use Only
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts