missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dynamitin (1-401 AA) Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 11361-1-AP-150UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
This antibody is against the full length p50 dynamitin.
This gene encodes a 50-kD subunit of dynactin, a macromolecular complex consisting of 10-11 subunits ranging in size from 22 to 150 kD. Dynactin binds to both microtubules and cytoplasmic dynein. It is involved in a diverse array of cellular functions, including ER-to-Golgi transport, the centripetal movement of lysosomes and endosomes, spindle formation, chromosome movement, nuclear positioning, and axonogenesis. This subunit is present in 4-5 copies per dynactin molecule. It contains three short alpha-helical coiled-coil domains that may mediate association with self or other dynactin subunits. It may interact directly with the largest subunit (p150) of dynactin and may affix p150 in place. Multiple alternatively spliced transcript variants encoding distinct isoforms have been found for this gene.Spezifikation
Dynamitin (1-401 AA) | |
Polyclonal | |
Unconjugated | |
DCTN2 | |
DCTN 50, DCTN2, DCTN50, dynactin 2, dynactin 2 (p50), Dynactin subunit 2, DYNAMITIN, p50 dynamitin, RBP50 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10540, 299850, 69654 | |
-20°C | |
Liquid |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
0.17 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q13561, Q6AYH5, Q99KJ8 | |
Dctn2 | |
Dynamitin (1-401 AA) Fusion Protein Ag1902 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |