missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DUX3 Monoclonal Antibody (3D2), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA524459
Additional Details : Gewicht : 0.01000kg
Beschreibung
Peptide Sequence: MALLTALDDT LPEEAQGPGR RMILLSTPSQ SDALRACFER NLYPGIATKE QLAQGIDIPE PRVQIWFQNE RSCQLRQHRR QSRPWPGRRD PQKGR
DUX3 Monoclonal antibody specifically detects DUX3 in Human samples. It is validated for ELISA, Western Blot.Spezifikation
DUX3 | |
Monoclonal | |
Unconjugated | |
DUX3 | |
double homeobox 3; double homeobox, 3; DUX3; putative double homeobox protein 3 | |
Mouse | |
Protein A | |
RUO | |
26582 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
3D2 | |
PBS with no preservative; pH 7.4 | |
Q96PT4 | |
DUX3 | |
DUX3 (NP_036280, 1 a.a. ∽ 95 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2b κ |