missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DUSP19 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 12924-1-AP-20UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
Dual-specificity phosphatases.Spezifikation
DUSP19 | |
Polyclonal | |
Unconjugated | |
DUSP19 | |
DUSP17, DUSP19, LMW DSP3, LMWDSP3, Protein phosphatase SKRP1, SKRP1, TS DSP1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
142679, 311151, 68082 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin) | |
0.23 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q8K4T5, Q8WTR2 | |
Dusp19 | |
DUSP19 Fusion Protein Ag3611 | |
20 μL | |
Primary | |
Rat, Mouse, Human | |
Antibody | |
IgG |