Learn More
DRY BLOCK HEATED AND STIRRING 25 X 24MM DIA.tubes temp range +5-150C stabilty 0.5C speed
Marke:  Bibby Scientific PS80025
DRY BLOCK HEATED AND STIRRING 25 X 24MM DIA.tubes temp range +5-150C stabilty 0.5C speed
Additional Details : Gewicht : 0.80000kg
Artikelnummer. 11767519