missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DRAM2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA563982
Additional Details : Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: HSGNFGTDLE QKLHWNPEDK GYVLHM Highest antigen sequence identity to the following orthologs: Mouse - 73%, Rat - 73%.
DRAM2 Polyclonal antibody specifically detects DRAM2 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin).Spezifikation
DRAM2 | |
Polyclonal | |
Unconjugated | |
DRAM2 | |
2010305N14Rik; 2610318G18Rik; AI647667; CORD21; damage regulated autophagy modulator 2; DNA damage regulated autophagy modulator 2; DNA damage-regulated autophagy modulator protein 2; DNA-damage regulated autophagy modulator 2; DRAM2; PRO180; PSEC0031; RP5-1180E21.1; TMEM77; Transmembrane protein 77; UNQ154/PRO180; VDNA-damage regulated autophagy modulator 2; WWFQ154 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
128338 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q6UX65 | |
DRAM2 | |
Recombinant protein corresponding to Human DRAM2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only