Learn More
Hach Lange™ DR 3900 Spectrophotometer with RFID technology
Description
- Sample preparation: reduced handling steps, precise dosing
- Sample analysis: automatic test recognition, expiration date check etc.
- Documentation of results: versatile data management, control cards
- With your daily routine of water analysis in mind the DR3900 spectrophotometer is optimised for safe processes and meaningful reports
- Control-parameters like Ammonium, COD, Phosphate, Nitrogen and many others are easy to perform
- The handling of tests and photometer is well designed to avoid any errors in your water analysis
Specifications
Specifications
| Höhe der Strahlmitte | 10 mm |
| Angaben zur Küvette | Compatibility: 10 mm, 50 mm, 1 in. rectangular and 13 mm, 1 in. round |
| Display | Touchscreen |
| Abmessungen (L x B x H) | 255 x 350 x 151 mm |
| Typ | Spektrophotometer zur Wasseranalyse |
| Luftfeuchtigkeit | 80 |
| Modell | DR3900 |
| Betriebstemperatur | 10°C to 40°C |
| Optische Konstruktion | Reference beam, spectral |