missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DOG-1 / TMEM16A Rabbit, Clone: DG1/1487R, enQuireBio™
Rabbit Monoclonal Antibody
Marke: EnquireBio 55107-RBM7-P1
Additional Details : Gewicht : 0.01000kg
Spezifikation
DOG-1 / TMEM16A | |
Cell Surface and Cytoplasm | |
DG1/1487R | |
Unconjugated | |
NP_011543428 | |
Rabbit | |
∽114kDa | |
Protein A/G Purified | |
RUO | |
55107 | |
IgG κ |
Immunohistochemistry | |
Monoclonal | |
200μg/mL | |
10mM PBS with or without 0.05% BSA and 0.05% azide. | |
TMEM16A | |
Recombinant full-length human DOG-1 protein | |
Gastrointestinal Stromal Tumor (GIST) or testicular germ cell tumor. Melanocytes in the basal layer of the epidermis and mast cells in the dermis of normal skin. | |
100 μg | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |