Learn More
DNAJC21 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA559300
Additional Details : Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: HPFYAYWQSF CTQKNFAWKE EYDTRQASNR WEKRAMEKEN KKIRDKARKE KNELVRQLVA FIRKRDKRVQ AHRKLVEEQN A Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 100%.
May act as a co-chaperone for HSP70. May play a role in ribosomal RNA (rRNA) biogenesis, possibly in the maturation of the 60S subunit. Binds the precursor 45S rRNA.Spezifikation
DNAJC21 | |
Polyclonal | |
Unconjugated | |
DNAJC21 | |
4930461P20Rik; 9930116P15Rik; DnaJ (Hsp40) homolog, subfamily C, member 21; DnaJ heat shock protein family (Hsp40) member C21; dnaJ homolog subfamily A member 5; DnaJ homolog subfamily C member 21; DnaJ homology subfamily A member 5; dnaja5; dnajc21; fa66c02; GS3; JJJ1; JJJ1 DnaJ domain protein homolog; Protein GS3; putative regulation protein GS3; Unknown (protein for MGC:63563); wu:fa66c02; zgc:63563 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
134218 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q5F1R6 | |
DNAJC21 | |
Recombinant protein corresponding to Human DNAJC21 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only