missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DNAJB7 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 18540-1-AP-20UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
DNAJB7 belongs to the evolutionarily conserved DNAJ/HSP40 family of proteins, which regulate molecular chaperone activity by stimulating ATPase activity. DNAJ proteins may have up to 3 distinct domains: a conserved 70-amino acid J domain, usually at the N terminus; a glycine/phenylalanine (G/F)-rich region; and a cysteine-rich domain containing 4 motifs resembling a zinc finger domain (Ohtsuka and Hata, 2000 ).Spezifikation
DNAJB7 | |
Polyclonal | |
Unconjugated | |
DNAJB7 | |
DJ5, DNAJB7, HSC3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
150353, 57755, 685839 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.33 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q7Z6W7, Q9QYI8 | |
Dnajb7 | |
DNAJB7 Fusion Protein Ag13367 | |
20 μL | |
Primary | |
Mouse, Human, Rat | |
Antibody | |
IgG |