missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dityrosine Antibody (10A6), Novus Biologicals™
Mouse Monoclonal Antibody
217.00 CHF - 715.00 CHF
Spezifikation
| Antigen | Dityrosine |
|---|---|
| Klon | 10A6 |
| Konzentration | 1 mg/mL |
| Verdünnung | Western Blot 1:100 - 1:2000, Flow Cytometry 1:50, ELISA 1:100 - 1:2000, Immunocytochemistry/Immunofluorescence 1:50 |
| Anwendungen | Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence |
| Product Code | Brand | Menge | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Menge | Price | Quantity & Availability | |||||
|
18246411
|
Novus Biologicals
NBP2-59361 |
100 μg |
715.00 CHF
100 Mikrogramm |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
|
18631627
|
Novus Biologicals
NBP2-59361-25ug |
25 μg |
217.00 CHF
25 Mikrogramm |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
Description
Dityrosine Monoclonal specifically detects Dityrosine in Non-species specific samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
| Dityrosine | |
| 1 mg/mL | |
| Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Synthetic Dityrosine conjugated to Keyhole Limpet Hemocyanin (KLH). | |
| Primary | |
| Specific for dityrosine modified proteins. Does not cross-react with 3,5-dibromotyrosine or bromotyrosine modified proteins. |
| 10A6 | |
| Western Blot 1:100 - 1:2000, Flow Cytometry 1:50, ELISA 1:100 - 1:2000, Immunocytochemistry/Immunofluorescence 1:50 | |
| Monoclonal | |
| Purified | |
| RUO | |
| IgG1 | |
| Protein G purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title