Learn More
BRAND™ Distillate Bottle for MonoDest 3000 E and N
Artikelnummer. 10059840 Alle ansehen Brand Produkte
Ansicht ändern
Click to view available options
Kapazität: 5 l
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer.
10059840 Kapazität: 5 l
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Artikelnummer. 10059840 Lieferant BRAND™ Lieferanten-Nr. 56300
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Alternative Produkte anzeigen
missing translation for 'validMainframeMsg'
Distillate bottle for MonoDest 3000 E and N is constructed of borosilicate glass.
  • Accessory for water still
TRUSTED_SUSTAINABILITY
Spezifikation
Produkttyp Distillate Bottle
Kapazität (metrisch) 5 L
Zur Verwendung mit (Geräte) MonoDest 3000 E and N
Material Borosilicate Glass