missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl(Phthalimidomethyl)phosphonat, 97 %, Thermo Scientific Chemicals

Catalog Number 11357865
Click to view available options
Menge:
2 g
10 g
50 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
28447-26-9
C11H12NO5P
269.193
MFCD00015464
WDBHXTMUHJGHAN-UHFFFAOYSA-N
dimethyl phthalimidomethyl phosphonate, dimethyl phthalimidomethylphosphonate, 2-dimethoxyphosphorylmethyl isoindole-1,3-dione, dimethyl 1,3-dioxoisoindolin-2-yl methyl phosphonate, dimethyl 1,3-dioxoisoindol-2-yl methylphosphonate, acmc-20al6l, dimethyl 1,3-dioxo-2,3-dihydro-1h-isoindol-2-yl methylphosphonate, dimethyl 1,3-dihydro-1,3-dioxo-2h-isoindol-2-yl methyl phosphonate, 1,3-dioxo-1,3-dihydro-isoindol-2-ylmethyl-phosphonic acid dimethyl ester, 1,3-dioxo-2,3-dihydro-1h-isoindole-2-ylmethyl phosphonic acid dimethyl ester
2773790
2-(Dimethoxyphosphorylmethyl)isoindol-1,3-dion
COP(=O)(CN1C(=O)C2=CC=CC=C2C1=O)OC
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
28447-26-9
C11H12NO5P
269.193
MFCD00015464
WDBHXTMUHJGHAN-UHFFFAOYSA-N
dimethyl phthalimidomethyl phosphonate, dimethyl phthalimidomethylphosphonate, 2-dimethoxyphosphorylmethyl isoindole-1,3-dione, dimethyl 1,3-dioxoisoindolin-2-yl methyl phosphonate, dimethyl 1,3-dioxoisoindol-2-yl methylphosphonate, acmc-20al6l, dimethyl 1,3-dioxo-2,3-dihydro-1h-isoindol-2-yl methylphosphonate, dimethyl 1,3-dihydro-1,3-dioxo-2h-isoindol-2-yl methyl phosphonate, 1,3-dioxo-1,3-dihydro-isoindol-2-ylmethyl-phosphonic acid dimethyl ester, 1,3-dioxo-2,3-dihydro-1h-isoindole-2-ylmethyl phosphonic acid dimethyl ester
2773790
2-(Dimethoxyphosphorylmethyl)isoindol-1,3-dion
COP(=O)(CN1C(=O)C2=CC=CC=C2C1=O)OC

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material Dimethyl (phthalimidomethyl)phosphonate
Schmelzpunkt 115°C to 120°C
Menge 2 g
Beilstein 1476370
Formelmasse 269.19
Reinheit (%) 97%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'