Learn More
ATAGO™ Digital pH Meter DPH-2
Click to view available options
Bereich (pH):
0 bis 14
Packungsgröße:
1 Stück
Beschreibung
- Easy to use
- Compact and portable
- IP67 water resistant
- Automatic Temperature Compensation mechanism that function from 0°C to 50°C.
- Applications include food and beverage production control, brewing and fermentation, soil and hydroponics, cutting oils, alkali washing liquids and other soluble solutions
Spezifikation
Spezifikation
| Genauigkeit (pH) | ± 0,1 |
| Zur Verwendung mit (Anwendung) | Food and Beverage Production Control, Brewing and Fermentation, Soil and Hydroponics, Cutting Oils, Alkali Washing Liquids, and Other Soluble Solutions |
| Bereich (pH) | 0 bis 14 |
| IP-Schutzart | IP67. |
| Modell | DPH-2 |
| Stromversorgung | LR44 Watch Battery (x4) |
| Auflösung | 0.1 pH |
| Temperaturkompensation | 0.0°C to 50.0°C |