missing translation for 'onlineSavingsMsg'
Learn More

Diethylenglykoldimethylether, 99 %, stab. mit100 ppm BHT, Thermo Scientific Chemicals

Catalog Number 11475403
Click to view available options
Menge:
500 mL
2500 mL
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Regulatorischer Hinweis: Dieses Produkt enthält Substanzen aus dem Anhang XIV der Verordnung REACH (EG) Nr. 1907/2006. Alle Kunden müssen vor der Lieferung eine Erklärung ausfüllen.
111-96-6
C6H14O3
MFCD00008503
SBZXBUIDTXKZTM-UHFFFAOYSA-N
diglyme, bis 2-methoxyethyl ether, 2-methoxyethyl ether, 1-methoxy-2-2-methoxyethoxy ethane, diethylene glycol dimethyl ether, poly-solv, di 2-methoxyethyl ether, dimethyl carbitol, 2,5,8-trioxanonane
8150
CHEBI:46784
1-Methoxy-2-(2-Methoxyethoxy)ethan
COCCOCCOC
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Regulatorischer Hinweis: Dieses Produkt enthält Substanzen aus dem Anhang XIV der Verordnung REACH (EG) Nr. 1907/2006. Alle Kunden müssen vor der Lieferung eine Erklärung ausfüllen.
111-96-6
C6H14O3
MFCD00008503
SBZXBUIDTXKZTM-UHFFFAOYSA-N
diglyme, bis 2-methoxyethyl ether, 2-methoxyethyl ether, 1-methoxy-2-2-methoxyethoxy ethane, diethylene glycol dimethyl ether, poly-solv, di 2-methoxyethyl ether, dimethyl carbitol, 2,5,8-trioxanonane
8150
CHEBI:46784
1-Methoxy-2-(2-Methoxyethoxy)ethan
COCCOCCOC

Diethylene glycol dimethyl ether is used as a solvent in organic reactions due to its stability towards higher pH and its high boiling point. It is particularly involved in reactions utilizing organometallic reagents such as Grignard reactions and metal hydride reductions. It is also a solvent for hydroboration reactions with diborane.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethylene glycol dimethyl ether is used as a solvent in organic reactions due to its stability towards higher pH and its high boiling point. It is particularly involved in reactions utilizing organometallic reagents such as Grignard reactions and metal hydride reductions. It is also a solvent for hydroboration reactions with diborane.

Solubility
Miscible with water, ethanol, acetone, acetic acid, glycerine, pyridine and aldehydes. Slightly miscible with ether.

Notes
Hygroscopic. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemischer Name oder Material Diethylene glycol dimethyl ether
Namenshinweis stabilized with 100ppm BHT
Schmelzpunkt -64°C
Dichte 0.944
Siedepunkt 161°C to 162°C
Flammpunkt 51°C (124°F)
Geruch Ether-like
Strukturformel (CH3OCH2CH2)2O
Brechungsindex 1.408
Menge 2500 mL
UN-Nummer UN3271
Beilstein 1736101
Empfindlichkeit Hygroskopisch
Merck Index 14,3165
Löslichkeitsinformationen Miscible with water,ethanol,acetone,acetic acid,glycerine,pyridine and aldehydes. Slightly miscible with ether.
Molekulargewicht (g/mol) 134.175
Formelmasse 134.18
Reinheit (%) 99%
Mehr anzeigen Weniger anzeigen

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'