missing translation for 'onlineSavingsMsg'
Learn More

Diethyl 5-Hydantoylphosphonat, 96 %

Catalog Number 11338056
Click to view available options
Menge:
1 g
5 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
95378-36-2
C7H13N2O5P
236.164
MFCD00171308
PNQYQNIOVAYJJG-UHFFFAOYSA-N
diethyl 2,5-dioxoimidazolidin-4-yl phosphonate, phosphonic acid, 2,5-dioxo-4-imidazolidinyl-, diethyl ester, diethyl 2,5-dioxoimidazolidin-4-ylphosphonate, 5-diethoxyphosphinyl hydantoin, diethyl 2,4-dioxoimidazoline-5-phosphonate, diethyl 2,4-dioxoimidazolidine-5-phosphonate, 5-diethoxycarbonyl-1,3-diazolidine-2,4-dione
3516088
5-Diethoxyphosphorylimidazolidin-2,4-dion
CCOP(=O)(C1C(=O)NC(=O)N1)OCC
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
95378-36-2
C7H13N2O5P
236.164
MFCD00171308
PNQYQNIOVAYJJG-UHFFFAOYSA-N
diethyl 2,5-dioxoimidazolidin-4-yl phosphonate, phosphonic acid, 2,5-dioxo-4-imidazolidinyl-, diethyl ester, diethyl 2,5-dioxoimidazolidin-4-ylphosphonate, 5-diethoxyphosphinyl hydantoin, diethyl 2,4-dioxoimidazoline-5-phosphonate, diethyl 2,4-dioxoimidazolidine-5-phosphonate, 5-diethoxycarbonyl-1,3-diazolidine-2,4-dione
3516088
5-Diethoxyphosphorylimidazolidin-2,4-dion
CCOP(=O)(C1C(=O)NC(=O)N1)OCC

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material Diethyl 5-hydantoylphosphonate
Schmelzpunkt 159°C to 161°C
Menge 1 g
Beilstein 4809527
Formelmasse 236.17
Reinheit (%) 96%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'