missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Di-Methyl-Lysine Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA596454
Additional Details : Gewicht : 0.01000kg
Beschreibung
Positive Samples: HeLa, H3 protein
Methylated lysine is derivative of the amino acid residue lysine where the sidechain ammonium group has been methylated one or more times.Spezifikation
Di-Methyl-Lysine | |
Polyclonal | |
Unconjugated | |
Dimethyllysine | |
A synthetic peptide corresponding to a sequence containing dimethylated K. | |
100 μL | |
Primary | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Dot Blot, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin) | |
1.62 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide, pH 7.3 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
Chemical, Human, Mouse, Rat | |
Antibody | |
IgG |