missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DHX36 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 13159-1-AP-20UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
Staufen is a member of the family of double-stranded RNA (dsRNA)-binding proteins involved in the transport and/or localization of mRNAs to different subcellular compartments and/or organelles. These proteins are characterized by the presence of multiple dsRNA-binding domains which are required to bind RNAs having double-stranded secondary structures. The human homologue of staufen encoded by STAU, in addition contains a microtubule- binding domain similar to that of microtubule-associated protein 1B, and binds tubulin. The STAU gene product has been shown to be present in the cytoplasm in association with the rough endoplasmic reticulum (RER), implicating this protein in the transport of mRNA via the microtubule network to the RER, the site of translation. Five transcript variants resulting from alternative splicing of STAU gene and encoding three isoforms have been described. Three of these variants encode the same isoform, however, differ in their 5′UTR.Spezifikation
DHX36 | |
Polyclonal | |
Unconjugated | |
DHX36 | |
DDX36, DEAH box protein 36, DHX36, G4R1, KIAA1488, MLE like protein 1, MLEL1, RHAU | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
170506, 310461, 72162 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin), Immunoprecipitation | |
0.19 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8VHK9, Q9H2U1 | |
Dhx36 | |
DHX36 Fusion Protein Ag3799 | |
20 μL | |
Primary | |
Rat, Human, Mouse | |
Antibody | |
IgG |