missing translation for 'onlineSavingsMsg'
Learn More

DGCR8 N-terminal Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

136.48 CHF - 385.92 CHF

Spezifikation

Antigen DGCR8 N-terminal
Konzentration 0.34 mg/mL
Anwendungen Immunocytochemistry, Western Blot, Immunofluorescence, Immunoprecipitation
Klassifikation Polyclonal
Konjugat Unconjugated
Mehr Spezifikationen ansehen

Produkte 2
Produktcode Marke Menge Preis Menge & Verfügbarkeit  
Produktcode Marke Menge Preis Menge & Verfügbarkeit  
16864395
Unterlagen ansehen
Proteintech
25835-1-AP-20UL
20 μL
136.48 CHF
20 Mikroliter
Alternative Produkte anzeigen
Sparen Sie bis zu 
»
om dit product te kopen Registreer vandaag om een webaccount aan te maken
16854395
Unterlagen ansehen
Proteintech
25835-1-AP-150UL
150 μL
385.92 CHF
150 Mikroliter
Alternative Produkte anzeigen
Sparen Sie bis zu 
»
om dit product te kopen Registreer vandaag om een webaccount aan te maken
Beschreibung

Beschreibung

This gene encodes a subunit of the microprocessor complex which mediates the biogenesis of microRNAs from the primary microRNA transcript. The encoded protein is a double-stranded RNA binding protein that functions as the non-catalytic subunit of the microprocessor complex. This protein is required for binding the double-stranded RNA substrate and facilitates cleavage of the RNA by the ribonuclease III protein, Drosha. Alternate splicing results in multiple transcript variants.
Spezifikation

Spezifikation

DGCR8 N-terminal
Immunocytochemistry, Western Blot, Immunofluorescence, Immunoprecipitation
Unconjugated
Rabbit
Human
Q8WYQ5
54487
DGCR8 N-terminal Fusion Protein Ag22988
Primary
-20°C
DGCR8
0.34 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
C22orf12, DGCR8, DGCRK6, Gy1
DGCR8
IgG
Antigen Affinity Chromatography
Antibody
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Product Certifications
Berichtigung von Produktinhalten

Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.

Name des Produkts

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.